Delmhorst 1849/24 Replacement Pins - 1-1/2" Insulated Pins
Representative image only.
Delmhorst 1849/24 Replacement Pins - 1-1/2" Insulated Pins
Delmhorst | Mfr: 1849/24| Item: 5982059
The wide assortment of replacement pins gives the flexibility to choose the right combination for your application
₹21,000.00INR / Each
₹21,000.00INR / Each
Product Description
Key Features
  • Configure your meter to your needs
  • 3 month warranty
More About this Item
Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..
24 per pack. Replacement pin for the 26-ES electrode
3 Month Warranty
Specifications
  • Measurement TypePin
  • Length (in)1 1/2
  • DescriptionReplacement Pins - 1-1/2" Insulated Pins