Decon Labs Proguard 9503 Professional Hand Cream, 3 oz
Representative image only.
Decon Labs Proguard 9503 Professional Hand Cream, 3 oz
Decon Labs | Mfr: 9503| Item: 8611526
Provides hands with protective barrier
₹1,800.00INR / Each
₹1,800.00INR / Each
Product Description
Key Features
  • Prevents irritation from latex gloves
  • Leave skin soft and protected
More About this Item
Specially formulated lotion moisturizes skin while providing a protective barrier that prevents irritation from latex gloves and other irritants. Does not contain grease, oil, perfume, aloe or other ingredients that can compromise the integrity of latex gloves. Absorbs quickly leaving skin soft and protected.
Specifications
  • FormatTube
  • DescriptionProfessional Hand Cream, 3 oz