Decon Labs Conflikt 4104 Detergent Disinfectant, 1 Gallon; 4/CS
Representative image only.
Decon Labs Conflikt 4104 Detergent Disinfectant, 1 Gallon; 4/CS
Decon Labs | Mfr: 4104| Item: 8611542
Ready-to-use disinfectant spray
₹37,000.00INR / Case of 4
₹37,000.00INR / Case of 4
Product Description
Key Features
  • Stable solution
  • USDA approved
  • EPA registered.
  • Refill bottles
More About this Item
This ready-to-use, prediluted quaternary ammonium disinfectant is ideal for cleaning and disinfecting hard surfaces in the lab, hospital, or just about anywhere that thorough disinfection efficacy is required. Effective against viruses, bacteria and fungi, HIV-1, E-coli and other pathogenic organisms including TB.
Not available for sale into California Not available for sale in Canada,
Specifications
  • FormatRefill
  • Volume (gallons)4
  • Volume (Liters)15
  • DescriptionDetergent Disinfectant, gallon, cs/4