Culture Bottle Roller Stacking Platform; Single
Representative image only.
Culture Bottle Roller Stacking Platform; Single
| Item: 3620252
Save lab counter space by stacking rollers
₹117,000.00INR / Each
₹117,000.00INR / Each
Buy It Together
Current Item
Culture Bottle Roller Stacking Platform; Single
Culture Bottle Roller Stacking Platform; Single
3620252
₹117,000.00INR / Each
+
Ibi Scientific Low-Profile Bottle Roller; 90-240 VAC, 50/60 Hz
3620203
₹1,099,000.00INR / Each
=
Total Price 1216000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Allows placement of additional roller inside incubator
  • Constructed of cast acrylic and aluminum
More About this Item
This raised platform is designed to fit inside an incubator to allow for stacking of one or two Low Profile rollers inside.
Constructed of cast acrylic and aluminum for years of service inside high humidity, CO2 incubators without discoloring or rusting.
Accommodates 1 additional roller unit to provide additional benchtop space, or to stack multiple units.
Specifications
  • MaterialCast Acrylic and Aluminum
  • Width (in)18
  • Height (in)9 3/8
  • Depth (in)16
  • DescriptionCulture Bottle Roller Stacking Platform; Single