CSL-843-075HC Inlet filter, polyester element, 99% efficient @ 5 micron, 25 CFM, 3/4" NPT(F)
Representative image only.
CSL-843-075HC Inlet filter, polyester element, 99% efficient @ 5 micron, 25 CFM, 3/4" NPT(F)
| Mfr: CSL-843-075HC| Item: 7977594
Protect your pump from dust and contaminants
₹16,300.00INR / Each
₹16,300.00INR / Each
Product Description
Key Features
  • 99% + efficient
  • vacuum levels of 1 x 10-3 mm Hg
More About this Item
Use inlet filters to protect your pump from any debris that may be in the air. The paper elements are disposable, replace when the element gets dirty. The polyester filter elements are reusable, simply clean off and reinstall. Replace element after three cleanings.
Specifications
  • MaterialPolyester element
  • DescriptionInlet filter, polyester element, 99% efficient @ 5 micron, 25 CFM, 3/4" NPT(F)