CSC Scientific 70535000 Upward Direction Tensiometer - +/-0.05 dyne/cm
Representative image only.
CSC Scientific 70535000 Upward Direction Tensiometer - +/-0.05 dyne/cm
CSC Scientific | Mfr: 70535000| Item: 5995120
Ideal for measuring the surface tension of oils, detergents, colloidal suspensions, and chemicals
Call For Price
for an additional
₹29,800.00INR / Each
for an additional
₹41,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
CSC Scientific 70535000 Upward Direction Tensiometer - +/-0.05 dyne/cm
CSC Scientific 70535000 Upward Direction Tensiometer - +/-0.05 dyne/cm
5995120
Call For Price
+
InnoCal NIST-Traceable Calibration; Penetrometers
1705032
₹29,800.00INR / Each
+ + =
Total Price 339800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
a 6-cm platinum-iridium measuring ring, three extra torsion wires, and a carrying case.
Key Features
  • Measure surface tension in 15 to 30 seconds
  • Direct-reading scale in dyne/cm
  • 1 year warranty
More About this Item
Measure both the surface and interfacial tension of liquids. Range is 0 to 90 dyne/cm. Measurement values are reproducible to within 0.05 dyne/cm. These meters are specified for determining interfacial tension of oil against water by the ring method in ASTM D971, solutions of surface-active agents in ASTM D1331, testing synthetic rubber lattices in ASTM D1417, and determining surface tension of industrial water/wastewater in ASTM D1590.
Model 59951-20 reads surface tension and upward interfacial tension. Features graduated dial readings in both directions from 0 to 90 dyne/cm and a double vernier. Both the scales and the corresponding verniers have graduations and figures in different colors to indicate whether the force is in an upward or downward direction.
1 Year Warranty
Specifications
  • Accuracy±0.05 dyne/cm
  • Range0 to 90 dyne/cm
  • DescriptionUpward Direction Tensiometer - +/-0.05 dyne/cm