CPC (Colder) Miniature Quick-Disconnect Fitting, Threaded Body, Polypropylene, Straight-Through, 1/8" NPT(M) ID; 1/Ea
Representative image only.
CPC (Colder) Miniature Quick-Disconnect Fitting, Threaded Body, Polypropylene, Straight-Through, 1/8" NPT(M) ID; 1/Ea
CPC (Colder) | Mfr: SMPT0212| Item: 1200634
No more kinked tubing—allows tubing to rotate freely
₹2,000.00INR / Each
₹2,000.00INR / Each
Product Description
Key Features
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected—preventing kinked tubing and accidental disconnection
More About this Item
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.
Specifications
  • Fitting MaterialPolypropylene
  • Valve MaterialNone
  • O-Ring MaterialEPDM
  • Sterilization MethodEthylene oxide (ETO) or gamma
  • Connection TypeQuick Disconnect x Hose Barb
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Quick-Disconnect Flow Size (in)3/32
  • ColorAlmond
  • Min Temperature (° F)32
  • Max Temperature (° F)180
  • Min Temperature (° C)0
  • Max Temperature (° C)82
  • Length (cm)2.1844
  • DescriptionMiniature Quick-Disconnect Fitting, Threaded Body, Polypropylene, Straight-Through, 1/8" NPT(M) ID; 1/Ea