CPC (Colder) AseptiQuik® STC II Quick-Disconnect Fitting, Genderless Sanitary Adapter with AseptiQuik G®, High-Temperature Polycarbonate, 3/4" x 3/4"; 1/Each

Representative image only.

CPC (Colder) AseptiQuik® STC II Quick-Disconnect Fitting, Genderless Sanitary Adapter with AseptiQuik G®, High-Temperature Polycarbonate, 3/4" x 3/4"; 1/Each

CPC (Colder) | Mfr: AQGSTC2330900HT| Item: 5011178

Genderless sterilizable connectors simplify process integration while maximizing flexibility

₹53,600.00INR / Each

₹53,600.00INR / Each
Product Description

Key Features

  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Robust construction provides reliable and repeatable fluid transfer
  • Innovative three-port steam design
  • Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP
  • Autoclave up to 130°C (266°F) or use gamma irradiation
  • 0.5" nominal flow path size
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • 3/4" x 3/4" sanitary connections

More About this Item

The AseptiQuik® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, simply unsnap and flip down the protective tab cover, align connectors with the pull tabs hanging down, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.

The fitting features a unique three-port design for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. Use the Steam-Thru II SIP connection to sterilize the fitting while connected to stainless steel equipment. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle following media transfer. The “steam off” function helps minimize cross-contamination risks associated with reusable components. The Steam-Thru II thumb latch helps secure valve position as well as offer visual indication of process stage. Select from 3/4" and 1-1/2" sanitary connections for attachment to the processing equipment. Connectors withstand temperatures up to 135°C (275°F) for 60 minutes during SIP.

All AseptiQuik connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinum-Cured Silicone
  • Sterilization MethodFull connector assembly: gamma irradiation up to 50 kGy or autoclave up to 130°C (266°F) for 60 minutes
    Steam-in-Place (SIP): up to 135°C (275°F) for 60 minutes
  • Connection TypeQuick Disconnect x Sanitary x Sanitary
  • Sanitary Clamp Size (Port 1) (InchShort)3/4
  • Sanitary Clamp Size (Port 2) (in)3/4
  • Flow PatternStraight
  • ColorWhite with white pull tab and amber tint
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)35
  • Fitting Length (in)8 1/4
  • Fitting Width (in)3 1/16
  • Fitting Length (mm)209.8
  • Fitting Width (mm)77.5
  • Regulatory ApprovalsNon-DEHP;RoHS;ADCF;FDA;USP;Non-Phthalate
  • DescriptionQuick-Disconnect Fitting, Genderless Sanitary Adapter with AseptiQuik G®, High-Temperature Polycarbonate, 3/4" x 3/4"; 1/Ea
Documents & Certificates
Reviews
Q&A