CPC (Colder) AseptiQuik® L Quick-Disconnect Fitting, High-Flow, Genderless Hose Barb, Polycarbonate, 3/4" ID; 1/Each

Representative image only.

CPC (Colder) AseptiQuik® L Quick-Disconnect Fitting, High-Flow, Genderless Hose Barb, Polycarbonate, 3/4" ID; 1/Each

CPC (Colder) | Mfr: AQL17012| Item: 5011181

High-flow genderless sterilizable connectors simplify process integration while maximizing flexibility

₹28,800.00INR / Each

₹28,800.00INR / Each
Product Description

Key Features

  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Large internal diameter for fast efficient fluid transfer
  • Robust construction provides reliable and repeatable fluid transfer
  • 1" nominal flow path size
  • 3/4" ID hose barb

More About this Item

The AseptiQuik® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, simply unsnap and flip down the protective tab cover, align connectors with the pull tabs hanging down, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.

All AseptiQuik connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinum-Cured Silicone
  • Sterilization MethodGamma irradiation up to 50 kGy
  • Connection TypeQuick Disconnect x Hose Barb
  • Barb Size / Tubing Id (Port 1) (mm)19.1
  • Barb Size / Tubing Id (Port 1) (in)3/4
  • Flow PatternStraight
  • ColorWhite with blue pull tab
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)60
  • Fitting Length (in)2 15/16
  • Fitting Width (in)4 1/4
  • Fitting Length (mm)74.9
  • Fitting Width (mm)107.2
  • Regulatory ApprovalsNon-Phthalate;FDA;RoHS;USP;Non-DEHP;ADCF
  • DescriptionQuick-Disconnect Fitting, High-Flow, Genderless Hose Barb, Polycarbonate, 3/4" ID; 1/Ea
Documents & Certificates
Reviews
Q&A