Costar Spin-X 8169 Centrifuge Tube Filters, Spinfilter Ny 0.22um 200/cs
Representative image only.
Costar Spin-X 8169 Centrifuge Tube Filters, Spinfilter Ny 0.22um 200/cs
Costar Spin-X | Mfr: 8169| Item: 0193738
Membrane-containing filter unit within a centrifuge tube
Call For Price
Call For Price
Product Description
Key Features
  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels
More About this Item
Ideal for HPLC sample preparation.
Specifications
  • MembraneNylon
  • Tube Size (mL)2
  • Working Volume (µL)500
  • Pore Size (µm)0.22
  • DescriptionCentrifuge Tube Filters, Spinfilter Ny 0.22um 200/cs