Costar Reagent Reservoir, Sterile, 50 mL; 200/Cs

Representative image only.

Costar Reagent Reservoir, Sterile, 50 mL; 200/Cs

Costar | Mfr: 4870| Item: 0790097

Draw samples into pipette easily

Call For Price

Call For Price
Product Description

Key Features

  • Disposable sterile reservoirs eliminate washing and reduce cross-contamination
  • Manufactured from virgin white polystyrene
  • 1 year warranty

More About this Item

Single-use reservoirs save you the hassle of washing and the problem of cross-contamination.

1 Year Warranty
Specifications
  • SterileYes
  • Max Volume (mL)50
  • Overall Length (InchShort)5.75
  • Number Of Channels1
  • Body MaterialPolystyrene
  • Color CodeWhite
  • Package Of5/Bag and 40 Bags/Case
  • DescriptionReagent Reservoir, Sterile, 50 mL; 200/Cs
  • Warranty1 year
Reviews
Q&A