Costar Microplate Storage Mat, 96-Well, EVA; 100/Cs
Representative image only.
Costar Microplate Storage Mat, 96-Well, EVA; 100/Cs
Costar | Mfr: 3080| Item: 0172922
Tightly seals microplate wells to prevents spills and well-to-well contamination
Call For Price
Call For Price
Product Description
Key Features
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • Can be applied manually or with our Storage Mat Applicator
More About this Item
Storage mat seals each well individually for long-term storage—virtually eliminates moisture loss. Use storage mats with standard microplates and 1.0-mL deep-well assay blocks.
Specifications
  • Number of Wells96
  • Well ShapeRound
  • Surface TreatmentNot Treated
  • SterileNo
  • Plate TypeSealing Mat
  • Plate ColorTranslucent
  • MaterialEthyl Vinyl Acetate
  • DescriptionMicroplate Storage Mat, 96-Well, EVA; 100/Cs