Corning Vented Cap for Sidearm Fittings, Polyethylene, 33 mm Thread, 0.2 µm Hydrophobic Membrane; 6/Cs
Representative image only.
Corning Vented Cap for Sidearm Fittings, Polyethylene, 33 mm Thread, 0.2 µm Hydrophobic Membrane; 6/Cs
Corning | Mfr: 3968| Item: 0183755
Optimize performance of your spinner flasks
Call For Price
Call For Price
Buy It Together
Current Item
Corning Vented Cap for Sidearm Fittings, Polyethylene, 33 mm Thread, 0.2 µm Hydrophobic Membrane; 6/Cs
Corning Vented Cap for Sidearm Fittings, Polyethylene, 33 mm Thread, 0.2 µm Hydrophobic Membrane; 6/Cs
0183755
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with the Corning® reusable spinner flasks
  • Allows gas exchange while minimizing risk of contamination
  • Sterilized by gamma irradiation
More About this Item
Choose from a wide range of accessories and replacement parts suitable for use with the Corning® reusable spinner flasks.
Vented cap has 0.2 µm pore nonwettable (hydrophobic) membrane sealed directly to the cap.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • MaterialPolyethylene
  • Closure33 mm Thread
  • Description33mm Polyethylene Vented Cap for Sidearm Fittings