Corning Stripwell™ Strip Ejector; 5/Cs
Representative image only.
Corning Stripwell™ Strip Ejector; 5/Cs
Corning | Mfr: 2578| Item: 1302052
Designed for use with Corning® Stripwell™ strips and plates
Call For Price
Call For Price
Product Description
Key Features
  • Constructed of durable polystyrene
More About this Item
These accessories are compatible with the Corning® Stripwell™ flat bottom strips and 96-well plates. The strip holder is the 96-well egg crate style holder for 1 x 8 Stripwell™ strips, model 13020-12. The strip ejector is designed to remove Stripwell™ strips from the egg crate holder. Strips and plates sold separately.
Specifications
  • Lid (Included)No
  • Plate TypeEjector
  • MaterialPolystyrene
  • Height (in)25/64
  • Width (in)4 7/64
  • Length (in)5 49/64
  • DescriptionStrip Ejector; 5/Cs