Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 5 mL; 200/Cs

Representative image only.

Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 5 mL; 200/Cs

Corning | Mfr: 4487| Item: 1300140

Bidirectional graduations provide choice of ascending and descending scales

Call For Price

Call For Price

Buy It Together

Current Item

Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 5 mL; 200/Cs
Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 5 mL; 200/Cs
1300140

Call For Price
+ =
Total Price 9300.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Negative graduations allow additional working volume
  • Calibrated "to deliver" for accurate dispensing to ±2%
  • Sterile, individually wrapped, nonpyrogenic, DNase-/RNase-free, and Human DNA-free

More About this Item

The Stripette™ serological pipettes are constructed of polystyrene and feature color-coded vertical stripes which are magnified by internal reagents, making the meniscus easy to read for quick volume identification. Uniform mouthpieces fit all standard pipette fillers. Individually wrapped paper/plastic packaging with "Banana peel" design make pipettes easy to open and remove aseptically with one hand, ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.

Uniform mouthpieces fit all standard pipette fillers. "Banana peel" (paper/plastic-wrap) packaging make them easy to open and remove pipettes aseptically with one hand.

Specifications
  • SterileYes
  • Max Volume (mL)5
  • Graduations (mL)0.1
  • Graduation IntervalNegative Graduation: 2.5 mL
  • Accuracy (%)±2%
  • Overall Length (InchShort)13.7
  • Overall Length (mm)348
  • Tip Inside Diameter (ID) (mm)1.85
  • Body MaterialPolystyrene
  • Color CodeBlue
  • CalibrationTo Deliver
  • Package OfIndividually Wrapped Paper/Plastic
  • DescriptionSerological Pipettes, Polystyrene, Sterile, 5 mL; 200/Cs
Reviews
Q&A