Corning Gosselin Autoclavable Bags, 9L, polypropylene, with biohazard symbol; 500/cs
Representative image only.
Corning Gosselin Autoclavable Bags, 9L, polypropylene, with biohazard symbol; 500/cs
Corning Gosselin | Mfr: SA09-10| Item: 3455194
Safely decontaminate waste in your autoclave
₹61,100.00INR / Case of 500
₹61,100.00INR / Case of 500
Product Description
Key Features
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are available with a biohazard logo
More About this Item
Gosselin autoclavable PP bags are 40 µm thick—strong enough to hold your lab waste during the autoclaving process. Bags withstand temperatures up to 273°F (134°C).
Specifications
  • Length (")12
  • MaterialPP
  • Volume (Liters)9
  • Height (InchShort)20
  • Thickness (mil)0.04