Corning CoolRack® V16 Cooling Block, Nine 16x100 mm Tubes; Gray
Representative image only.
Corning CoolRack® V16 Cooling Block, Nine 16x100 mm Tubes; Gray
Corning | Mfr: 432066| Item: 0439266
Cool, freeze, heat, or thaw samples uniformly
₹128,000.00INR / Each
₹128,000.00INR / Each
Buy It Together
Current Item
Corning CoolRack® V16 Cooling Block, Nine 16x100 mm Tubes; Gray
Corning CoolRack® V16 Cooling Block, Nine 16x100 mm Tubes; Gray
0439266
₹128,000.00INR / Each
+ =
Total Price 146600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
  • 1 year warranty
More About this Item
These aluminum blocks provide better temperature uniformity than placing samples directly into a bath. Aluminum blocks may be autoclaved, high-heat sterilized, or chemically treated to decrease cross-contamination concerns. Safe for use in ice, dry ice, alcohol, liquid nitrogen, water, oil, and other temperature regulation baths. Use in traditional ice buckets and pans, or use with a Corning CoolBox™ for an ice-free option.
1 Year Warranty
Specifications
  • Tubes Held9
  • Tube Size16 x 100 mm tubes
  • ColorGray
  • Capacity9 x 16x100 mm tubes
  • MaterialAluminum
  • DescriptionCooling Block, Nine 16x100 mm Tubes; Gray