Corning 431175 45 mm Neck Polystyrene Storage Bottles, 150 mL; 24/Cs

Representative image only.

Corning 431175 45 mm Neck Polystyrene Storage Bottles, 150 mL; 24/Cs

Corning | Mfr: 431175| Item: 2953102

The new low profile design with easy-grip handles facilitate handling

Call For Price

Call For Price
Product Description

Key Features

  • Plug-seal cap provides airtight seal and helps minimize the risk of contamination
  • Sterile

More About this Item

Store media, buffers, and other aqueous solutions in these disposable polystyrene bottles. Replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

Specifications
  • Capacity (mL)150
  • Capacity (oz)5
  • MaterialPolystyrene
  • Cap MaterialHigh-Density Polyethyelene (HDPE)
  • Cap / Stopper Size45 mm
  • Cap ColorOrange
  • Description45 mm Neck PS Storage Bottles, 150 mL; 24/Cs
Reviews
Q&A