Corning 430282 45 mm Neck Polystyrene Storage Bottles, 500 mL; 24/Cs

Representative image only.
Corning Mfr # 430282 – Item # WW-29531-06
The new low profile design with easy-grip handles facilitate handling
  • Plug-seal cap provides airtight seal and helps minimize the risk of contamination
  • Sterile

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialPolystyrene
  • Cap MaterialHigh-Density Polyethyelene (HDPE)
  • Cap / Stopper Size45 mm
  • Cap ColorOrange
  • Description45 mm Neck PS Storage Bottles, 500 mL; 24/Cs

Key Features

  • Plug-seal cap provides airtight seal and helps minimize the risk of contamination
  • Sterile

More About this Item

Store media, buffers, and other aqueous solutions in these disposable polystyrene bottles. Replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat