Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, purple, 1/ea
Representative image only.
Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, purple, 1/ea
Corning | Mfr: 432025| Item: 0439225
No mess, no maintenance
₹345,000.00INR / Each
₹345,000.00INR / Each
Buy It Together
Current Item
Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, purple, 1/ea
Corning 2XT CoolBox™ Ice-Free Cooler with Two Cooling Cores, purple, 1/ea
0439225
₹345,000.00INR / Each
+ =
Total Price 372500.00 ADD 2 ITEMS TO CART
Product Description
Includes
Cooling cores and removable lid.
Key Features
  • Keep samples cold without melting ice contamination, inconvenient batteries, or power cord clutter
  • Cooling core maintains sample temperatures at 0.5°C to 4°C for at least four hours
  • Compatible with all Corning CoolSink® and CoolRack® sample modules (order separately)
  • 1 year warranty
More About this Item
These benchtop coolers eliminate possible contamination from ice, maintenance costs associated with battery replacement, and messy cords. Simply freeze the cooling core in a -20°C freezer for 12 or more hours, then remove and wait for core to reach 1°C before placing in holder. Once the core is installed, add your CoolSink or CoolRack module and load your samples. Samples quickly equilibrate to the core temperature and remain chilled for 4 to 16 hours.
Select the CoolBox system for longer-term cooling, which features a deeper sample area capable of maintaining temperature up to 16 hours.
1 Year Warranty
Specifications
  • Lowest Set Temperature (° C)-20
  • ColorPurple
  • Width (in)10 7/16
  • Width (cm)26.5
  • DescriptionModule with Two Cooling Cores, purple, 1/ea