Corning 2158-500 Concentrator with Springs, 500 Ml
Representative image only.
Corning 2158-500 Concentrator with Springs, 500 Ml
Corning | Mfr: 2158-500| Item: 3453000
Efficiently boil away pestistides for accurate testing
₹122,000.00INR / Each
₹122,000.00INR / Each
Product Description
Key Features
  • Held together using glass hooks and two 1 1/4 inch stainless steel springs
More About this Item
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available with springs mounted on collection tube.
Specifications
  • Accessory TypeConcentrator
  • Range500 mL
  • DescriptionConcentrator with Springs, 500 Ml