Cooper-Atkins 52048-K Refrigerator/Freezer Thermocouple; Type K Solid Simulator
Representative image only.
Cooper-Atkins 52048-K Refrigerator/Freezer Thermocouple; Type K Solid Simulator
Cooper-Atkins | Mfr: 52048-K| Item: 9002519
₹20,000.00INR / Each
₹20,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
The Thermocouple Solid SImulator does not measure the air, it takes on refrigeration temperature as food does and further supports your food safety program. Made of FDA compliant material it simulates food product temperature in changing ambient conditions of any refrigerator, freezer, walk-in or delivery truck. The temperature of the Solid Simulator will display immediately when plugged into any Type K Thermocouple Instrument. Walk In...And Plug In!
1 Year Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Sheath Length (in)1.5
  • Sheath Diameter (InchShort)1.5
  • DescriptionRefrigerator/Freezer Thermocouple; Type K Solid Simulator