Contec Prosat® PS-911 Presaturated Cleanroom Wipes; 30 Wipes/Pack, 10 Pks/Cs
Representative image only.
Contec Prosat® PS-911 Presaturated Cleanroom Wipes; 30 Wipes/Pack, 10 Pks/Cs
Contec | Mfr: PS-911, PROSAT| Item: 3367500
Ideal for general-purpose cleaning in Class 100 environments
Call For Price
Call For Price
Product Description
More About this Item
Prosat wipes measure 9" x 11" and are presaturated in a solvent blend of 30% DI water and 70% isopropyl alcohol; solvents are filtered to a 0.2 micron level. Low-lint polypropylene wipes won’t scratch delicate surfaces. Wipes come in a convenient pack of 30 that can be easily ransported and used gain. Wipes are sold in cases of 10 packs of 30 wipes/pk.
Note: Not available for sale to the state of California
Specifications
  • ComplianceClass 100
  • DescriptionPresaturated cleanroom wipes, 300/pack