Contec MicroGenesis Microfiber Wipe, 100/bag
Representative image only.
Contec MicroGenesis Microfiber Wipe, 100/bag
Contec | Mfr: LWMM0100| Item: 3367515
Multi-filament polyester microfiber gives excellent pick up and retention of particles
₹14,500.00INR / Bag of 100
₹14,500.00INR / Bag of 100
Product Description
Key Features
  • Ideal for picking up powder residue created during pharmaceutical manufacturing
More About this Item
The MicroGenesis wipe is constructed of 100% polyester microfiber yarn knitted into a thermally-bonded polypropylene substrate, delivering the maximum amount of microfiber to pick up and retain particles. Use this wipe while manufacturing tablet and fine-powdered pharmaceuticals to pick up powder residue, remove grease, clean glasses and goggles, and with disinfectants; for general cleanrooms, use for spill pick up, wet and dry floor cleaning, and stainless steel cleaning and polishing.
Specifications
  • MaterialPolyester
  • Length (InchShort)12
  • Width (InchShort)12
  • Width (cm)31
  • Length (cm)31
  • DescriptionMicrofiber Wipe, 100/bag