Contec EasyCurve Mop Frame, stainless steel, autoclavable
Representative image only.
Contec EasyCurve Mop Frame, stainless steel, autoclavable
Contec | Mfr: 2724| Item: 0941141
Curved mop face with pivoting connector provides excellent surface contact and maneuverability
₹30,800.00INR / Each
₹30,800.00INR / Each
Product Description
Key Features
  • The lighweight and maneuverability of the stainless steel mop makes it perfect for cleaning floors, walls, and ceilings
More About this Item
Designed for use in controlled environments. Curved head and pivoting connector ensures proper lift and pull cleaning technique. Autoclavable.
Polyurethane foam mop head has a laminated fabric face for durability. Polypropylene mop bucket features a sling-style wringer that effectively wrings the mop and removes contaminants from the mop head. Twin-divided bucket holds up to 5.3 gallons (20 L) and includes a cover, sling wringer, and casters. All parts are autoclavable except the casters. Mop head and bucket are sold separately.
ISO9001 Certified Supplier
Specifications
  • MaterialStainless steel
  • Width (in)14
  • Length (in)60
  • Depth (in)4.25
  • DescriptionMop Frame, stainless steel, autoclavable