Compression Fitting, Reducing Union, PP, 1/2" OD x 3/8" OD; 10/Pk
Representative image only.
Compression Fitting, Reducing Union, PP, 1/2" OD x 3/8" OD; 10/Pk
| Item: 3623474
For water-treatment, DI water, and mild chemical-feed tubing connections
₹5,800.00INR / Pkg of 10
₹5,800.00INR / Pkg of 10
Product Description
Key Features
  • 20% glass-filled polypropylene offers good chemical resistance and improved strength
  • Ideal for water-treatment, filtration, DI water, and mild chemical-feed connections
  • Unaffected by alkalis, salts, and most weak acids
  • Material temperature rating: -34 to 107°C (-30 to 225°F)
  • Plastic gripper nut helps secure plastic tubing
  • NSF 14 certified, NSF 61 compliant, and autoclavable
  • 1/2" OD x 3/8" OD
More About this Item
JACO polypropylene compression reducing union fittings connect rigid plastic tubing to equipment ports in water-treatment, filtration, deionized-water, and mild chemical-feed systems. Molded from 20% glass-filled polypropylene, these fittings are lightweight and rigid, with improved strength, resistance to environmental stress cracking, and good chemical resistance.
PVDF provides strong durability, aging resistance, and chemical compatibility across a broad working range, making it a dependable choice for pharmaceutical, biotech, and food and beverage process-support applications where PVDF compatibility and required certifications have been verified.
These fittings have a semi-translucent, cloudy off-white appearance. PVDF complies with FDA food-contact requirements under 21 CFR § 177.2510, is NSF Listed, and autoclavable.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPP
  • Connection TypeCompression
  • Number Of Ports2
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Compression Size / Tube Od (Port 2) (in)3/8
  • Flow PatternStraight
  • ColorNatural
  • Min Temperature (° F)-30
  • Max Temperature (° F)225
  • Min Temperature (° C)-34
  • Max Temperature (° C)107
  • Max Operating Pressure (PSI)220
  • Quantity10
  • DescriptionCompression Fitting, Reducing Union, PP, 1/2" OD x 3/8" OD; 10/Pk