COMARK UTL80 Stick-On Dial Refrigerator/Freezer HACCP-Series Thermometer
Representative image only.
COMARK UTL80 Stick-On Dial Refrigerator/Freezer HACCP-Series Thermometer
COMARK | Mfr: UTL80| Item: 9002581
Use to check your refrigerator or freezer is at the correct temperature, keeping food safe to eat.
₹1,400.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,400.00INR / Each
Buy It Together
Current Item
COMARK UTL80 Stick-On Dial Refrigerator/Freezer HACCP-Series Thermometer
COMARK UTL80 Stick-On Dial Refrigerator/Freezer HACCP-Series Thermometer
9002581
₹1,400.00INR / Each
+ =
Total Price 20100.00 ADD 2 ITEMS TO CART
Product Description
Includes
double stick tape
Key Features
  • Accuracy: ±1 division
  • Division 2°
  • Easy-to-read color HACCP zones
  • NSF Approved
  • 1 year warranty
More About this Item
The Comark UTL80 thermometer with colored temperature zones is designed for quick reading inside a refrigerator, freezer or cooler. The double stick tape allows for securing inside a refrigerator, freezer or cooler.
1 Year Warranty
Specifications
  • Min Temperature (° F)-20
  • Min Temperature (° C)-30
  • Max Temperature (° F)80
  • Max Temperature (° C)25
  • Division (° C)2
  • Division (° F)2
  • DescriptionStick-On Dial Refrigerator/Freezer HACCP-Series Thermometer