COMARK FWT Freezer Wall Thermomter with HACCP-Series Zones, -10 to 80F
Representative image only.
COMARK FWT Freezer Wall Thermomter with HACCP-Series Zones, -10 to 80F
COMARK | Mfr: FWT| Item: 9002580
Designed specifically for use in the food industry but can be used in other applications also
₹3,600.00INR / Each
₹3,600.00INR / Each
Product Description
Key Features
  • Includes: bracket and mounting screws.
  • 1 year warranty
More About this Item
The Comark FWT is used to verify that refrigerated and frozen foods are being storred at suitable temperatures, in compliance with your HACCP plan. With a large, prominent display and clearly marked caution and danger zones, you can make your checks with a quick glance.
1 Year Warranty
Specifications
  • Min Temperature (° F)-10
  • Max Temperature (° F)80
  • Division (° F)2
  • DescriptionFreezer Wall Thermomter with HACCP-Series Zones, -10 to 80F