Cole-Parmer Wide-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK

Representative image only.

Cole-Parmer Wide-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK

Cole-Parmer | Item: 6215030

Transport hazardous samples reliably

₹12,200.00INR / Pkg of 6

₹12,200.00INR / Pkg of 6

Buy It Together

Current Item

Cole-Parmer Wide-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK
Cole-Parmer Wide-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK
6215030

₹12,200.00INR / Pkg of 6
+
Cole-Parmer Replacement Cap, PP, 63mm; 12/PK
6300014

₹7,700.00INR / Pkg of 12
=
Total Price 19900.00 ADD 2 ITEMS TO CART
Product Description

Includes

Polypropylene screw closures.

Key Features

  • Safe for small quantities of corrosives or cleaning supplies
  • Leakproof polypropylene screw cap

More About this Item

Safe, durable, and cost-efficient, these bottles allow you to transport solutions safety within your lab or facility. Wide mouth allows easy filling with dry materials or liquids. Designed to resist splitting and puncturing from shipping shocks.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

Specifications
  • Capacity (mL)1000
  • Capacity (L)1
  • Capacity (oz)32
  • Capacity (Gal)0.264200792602378
  • MaterialPlastic
  • Cap MaterialPolypropylene
  • Cap / Stopper Size63
  • Cap Colornatural
  • Bottle Colornatural
  • AutoclavableNo
  • Neck Inside Diameter (ID) (mm)52
  • Neck Inside Diameter (ID) (in)2 3/64
  • Outside Diameter (OD) (in)3 35/64
  • Outside Diameter (OD) (cm)9
  • Overall Height (in)7 23/32
  • Overall Height (cm)19.6
  • Overall Depth (in)3 35/64
  • Overall Depth (cm)9
  • Overall Width (in)3 35/64
  • Overall Width (cm)9
  • DescriptionWide-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK
Reviews
Q&A