Cole-Parmer Wide-Mouth Plastic Bottle, PFA, 500mL

Representative image only.

Cole-Parmer Wide-Mouth Plastic Bottle, PFA, 500mL

Cole-Parmer | Item: 0605296

Provides safe storage of corrosive and hazardous chemicals

₹16,900.00INR / Each

₹16,900.00INR / Each

Buy It Together

Current Item

Cole-Parmer Wide-Mouth Plastic Bottle, PFA, 500mL
Cole-Parmer Wide-Mouth Plastic Bottle, PFA, 500mL
0605296

₹16,900.00INR / Each
+
Cole-Parmer Narrow-Mouth Plastic Bottle, PFA, 100mL
0605291

₹7,500.00INR / Each
=
Total Price 24400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Leakproof PFA cap.

Key Features

  • Withstands temperatures from -200℃ to 260℃ (-328° to 500°F)
  • Chemical and corrosion resistance
  • Smooth inner and outer surface for easy cleaning
  • Wide mouth makes filling and pouring easy
  • Autoclavable
  • 6 month warranty

More About this Item

These perfluoroalkoxy (PFA) wide-mouth bottles offer excellent resistance to chemicals, high and low temperatures, and corrosion. Recommended for demanding fluid handling and storage applications. Safe for reagents which can’t be stored in metal containers.

The translucent bottles can also be used for solutions or chemicals commonly used for trace quantity analysis, ultra-trace quantity analysis, ICP-MS analysis, and isotopic analysis. They feature extremely low water-absorption, leakproof PFA caps, and non-stick surface for easy cleaning.

Before autoclaving, place cap onto bottle mouth, but do not tighten.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

6 Month Warranty
Specifications
  • Capacity (mL)500
  • Capacity (L)0.5
  • Capacity (oz)16.9
  • Capacity (Gal)0.132
  • MaterialPerfluoroalkoxy (PFA)
  • Cap MaterialPFA
  • Cap / Stopper SizeGL45
  • Cap Colornatural
  • Bottle ColorNatural
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)500
  • Withstands Low Temperature (° F)-328
  • Neck Inside Diameter (ID) (mm)36
  • Neck Inside Diameter (ID) (in)1 27/64
  • Outside Diameter (OD) (in)3 5/64
  • Outside Diameter (OD) (cm)7.8
  • Overall Height (in)6 3/8
  • Overall Height (cm)16.2
  • Overall Depth (in)6 19/64
  • Overall Depth (cm)16
  • Overall Width (in)3 5/64
  • Overall Width (cm)7.8
  • DescriptionWide-Mouth Plastic Bottle, PFA, 500mL
  • Warranty6 month
Reviews
Q&A