Cole-Parmer Vortex Mixer Rack, 50 x 12 mm Tubes; 1/Ea
Representative image only.
Cole-Parmer Vortex Mixer Rack, 50 x 12 mm Tubes; 1/Ea
Cole-Parmer | Item: 0472562
Pre-drilled foam racks to meet your mixing needs
₹11,000.00INR / Each
₹11,000.00INR / Each
Product Description
Key Features
  • Use with Cole-Parmer High-Capacity Vortex Mixer/Shaker—standard or QuEChERS versions.
  • 2 year warranty
More About this Item
Lightweight foam racks can be used to hold samples vertically or horizontally.
Two year warranty
Specifications
  • MaterialEthyl vinyl acetate foam
  • Compatible With50 x 12 mm tubes
  • DescriptionVortex Mixer Rack, 50 x 12 mm Tubes; 1/Ea
  • Warranty2 Years