Cole-Parmer Vortex Mixer Rack, 3 x Plates/96-Well Microtube Racks; 1/Ea
Representative image only.
Cole-Parmer Vortex Mixer Rack, 3 x Plates/96-Well Microtube Racks; 1/Ea
Cole-Parmer | Item: 0472569
Pre-drilled foam racks to meet your mixing needs
₹42,400.00INR / Each
₹42,400.00INR / Each
Product Description
Key Features
  • Use with Cole-Parmer High-Capacity Vortex Mixer/Shaker—standard or QuEChERS versions.
  • 2 year warranty
More About this Item
Lightweight foam racks can be used to hold samples vertically or horizontally.
Two year warranty
Specifications
  • MaterialPolypropylene
  • Compatible With3 x plates/96-well microtube racks
  • DescriptionVortex Mixer Rack, 3 x Plates/96-Well Microtube Racks; 1/Ea
  • Warranty2 Years