Cole-Parmer Vortamix™ Mini Vortexer Gasket Cap, Blue; 1/Ea
Representative image only.
Cole-Parmer Vortamix™ Mini Vortexer Gasket Cap, Blue; 1/Ea
Cole-Parmer | Item: 0439730
Order the replacement red gasket cap; or blue, green, or purple caps to fit your preference
₹1,600.00INR / Each
₹1,600.00INR / Each
Product Description
Key Features
  • Construced of durable rubber
More About this Item
The standard unit comes with a red rubber gasket cap, but blue, green and purple colored caps are also available. Vortamix mini vortexer sold separately as model 04397-27.
The innovative design of the gasket cap allows centrifuge and test tubes up to 30 mm in diameter to be mixed with very little effort – including 50mL conical tubes!
Specifications
  • ColorBlue
  • Compatible WithArgos Vortamix vortex shaker
  • DescriptionMini Vortexer Gasket Cap, Blue; 1/Ea