Cole-Parmer VapLock™ Solvent Waste Manifold, PTFE, panel mount, 1x1/2"NPT, 3x1/4"NPT, 6x1/4-28; 1/ea
Representative image only.
Cole-Parmer VapLock™ Solvent Waste Manifold, PTFE, panel mount, 1x1/2"NPT, 3x1/4"NPT, 6x1/4-28; 1/ea
Cole-Parmer | Item: 1201975
Connect solvent waste manifold caps to waste containers for a secure closed system
₹42,800.00INR / Each
₹42,800.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Solvent Waste Manifold, PTFE, panel mount, 1x1/2"NPT, 3x1/4"NPT, 6x1/4-28; 1/ea
Cole-Parmer VapLock™ Solvent Waste Manifold, PTFE, panel mount, 1x1/2"NPT, 3x1/4"NPT, 6x1/4-28; 1/ea
1201975
₹42,800.00INR / Each
+ =
Total Price 49800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Choose from polypropylene or PTFE construction
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Panel-mount caps fit a 1-1/4" ID opening with panel thickness from 1/16" to 1/2" and comes with PP or PTFE locking nut. A 1-3/4" diameter obstruction-free area around the opening is required for use. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
Panel-mount waste caps are configured to fit an aperture drilled to 1-1/4" ID with panel thickness of 1/16" to 1/2". An area 1-3/4" dia free of obstruction around the aperture is required for use. Manifold drains to a 1/2" NPT common port at the base.
Specifications
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap / Stopper SizePanel Mount
  • Special Features1x1/2"NPT, 3x1/4"NPT, 6x1/4-28
  • DescriptionSolvent Waste Manifold, PTFE, panel mount, 1x1/2"NPT, 3x1/4"NPT, 6x1/4-28; 1/ea