Cole-Parmer VapLock™ Solvent Waste Manifold, PP, Justrite SS, 1x1/2" NPTF, 3x1/4" NPTF; 1/ea
Representative image only.
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, Justrite SS, 1x1/2" NPTF, 3x1/4" NPTF; 1/ea
Cole-Parmer | Item: 1201958
Connect solvent waste manifold caps to waste containers for a secure closed system
₹26,500.00INR / Each
₹26,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, Justrite SS, 1x1/2" NPTF, 3x1/4" NPTF; 1/ea
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, Justrite SS, 1x1/2" NPTF, 3x1/4" NPTF; 1/ea
1201958
₹26,500.00INR / Each
+ + =
Total Price 44600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from polypropylene or PTFE construction
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Quick disconnect caps are available with PP or SS fittings and allow fast and easy connections to Justrite® safety containers. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
Specifications
  • MaterialPolypropylene(PP)
  • Cap / Stopper SizeJustrite Quick-Disconnect, SS
  • Special Features1x1/2" NPTF, 3x1/4" NPTF
  • DescriptionSolvent Waste Manifold, PP, Justrite SS, 1x1/2" NPTF, 3x1/4" NPTF; 1/ea