Cole-Parmer VapLock™ Solvent Delivery Cap w/ EPDM Air Inlet Valve, Two 1/4-28 ports, 38 mm Wako; 1/EA

Representative image only.

Cole-Parmer VapLock™ Solvent Delivery Cap w/ EPDM Air Inlet Valve, Two 1/4-28 ports, 38 mm Wako; 1/EA

Cole-Parmer | Item: 1201842

Reduce chemical evaporation and exposure to hazardous vapors

₹6,200.00INR / Each

₹6,200.00INR / Each

Buy It Together

Current Item

Cole-Parmer VapLock™ Solvent Delivery Cap w/ EPDM Air Inlet Valve, Two 1/4-28 ports, 38 mm Wako; 1/EA
Cole-Parmer VapLock™ Solvent Delivery Cap w/ EPDM Air Inlet Valve, Two 1/4-28 ports, 38 mm Wako; 1/EA
1201842

₹6,200.00INR / Each
+ + + + =
Total Price 123970.00 ADD 5 ITEMS TO CART
Product Description

Key Features

  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
  • 38 mm Wako thread with two 1/4-28 ports
  • EPDM air inlet valve

More About this Item

Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.

Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 µm PTFE filter to reduce particulate contamination. Select from FFKM, EPDM, or Viton® valve seal options.

*Does not come in contact with liquid under normal circumstances.

Specifications
  • Cap MaterialEPDM® air inlet valve
  • Cap / Stopper SizeWako 38 mm
  • Cap ColorWhite
  • AutoclavableYes
  • Special Features2 x 1/4"-28 UNF(F)
  • DescriptionSolvent Delivery Cap w/ EPDM Air Inlet Valve, two 1/4"-28 ports, 38 mm Wako; 1/ea
Reviews
Q&A