Cole-Parmer VapLock™ Port Plug, Polypropylene, 1/4" NPT(M); 10/PK
Representative image only.
Cole-Parmer VapLock™ Port Plug, Polypropylene, 1/4" NPT(M); 10/PK
Cole-Parmer | Item: 1202044
Designed for secure leak-resistant connections
₹1,600.00INR / Pkg of 10
₹1,600.00INR / Pkg of 10
Product Description
Key Features
  • Available in multiple materials and sizes
  • Wrap with PTFE tape for leak-resistant connections
More About this Item
Use plugs to seal unused manifold and bottle cap ports for a secure leak-resistant system. Options for NPT ports are 1/8", 1/4", 3/8", and 1/2" NPT. To ensure a leak-resistant connection, the plugs should be wrapped three to four times with PTFE tape.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeNPT(M)
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • ColorGray
  • DescriptionPort Plug, 1/4" NPT(M), Polypropylene; 10/pk