Cole-Parmer VapLock™ Fitting, Polypropylene, Tee, Hose Barb Union, 1/8" ID; 1/EA
Representative image only.
Cole-Parmer VapLock™ Fitting, Polypropylene, Tee, Hose Barb Union, 1/8" ID; 1/EA
Cole-Parmer | Item: 1202376
Connect soft tubing using barbed fittings
₹320.00INR / Each
₹320.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Fitting, Polypropylene, Tee, Hose Barb Union, 1/8" ID; 1/EA
Cole-Parmer VapLock™ Fitting, Polypropylene, Tee, Hose Barb Union, 1/8" ID; 1/EA
1202376
₹320.00INR / Each
+ =
Total Price 2020.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
More About this Item
These tee barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeBarb
  • Number Of Ports3
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Barb Size / Tubing Id (Port 3) (in)1/8
  • DescriptionFitting, Polypropylene, Tee, Hose Barb Union, 1/8" ID; 1/EA