Cole-Parmer VapLock™ Filter Adapter, 1/8" NPT(M) x 1/4" NPT(M), polypropylene; 1/ea
Representative image only.
Cole-Parmer VapLock™ Filter Adapter, 1/8" NPT(M) x 1/4" NPT(M), polypropylene; 1/ea
Cole-Parmer | Item: 1202000
Various adapter configurations allow quick and easy connection to emission sources
₹300.00INR / Each
₹300.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Filter Adapter, 1/8" NPT(M) x 1/4" NPT(M), polypropylene; 1/ea
Cole-Parmer VapLock™ Filter Adapter, 1/8" NPT(M) x 1/4" NPT(M), polypropylene; 1/ea
1202000
₹300.00INR / Each
+ =
Total Price 7300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from polypropylene or nylon construction
More About this Item
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. These adapters are NPT(M) x NPT(M), including options for different sizes and choices between straight or elbow.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeNPT(M) x NPT(M)
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • ColorGray
  • DescriptionFilter Adapter, 1/8" NPT(M) x 1/4" NPT(M), polypropylene; 1/ea