Cole-Parmer Ultrasonic Processor Cup Horn Microtube Holder
Representative image only.
Cole-Parmer Ultrasonic Processor Cup Horn Microtube Holder
Cole-Parmer | Item: 0471043
Process a sample in isolation from the probe
₹12,300.00INR / Each
₹12,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer Ultrasonic Processor Cup Horn Microtube Holder
Cole-Parmer Ultrasonic Processor Cup Horn Microtube Holder
0471043
₹12,300.00INR / Each
+ =
Total Price 12300.00 ADD 2 ITEMS TO CART
Product Description
Includes
vial holder and splash shield.
Key Features
  • Cooling water can be circulated through the cup during operation
  • 3 year warranty
More About this Item
Use this cup horn to process a sample in absolute isolation from the probe. The horn is especially useful for processing samples that must be kept in a sealed vessel, such as pathogens. The ultrasonic vibrations are transmitted through the water in the cup causing cavitation in the sample vessel.
3 Year Warranty
Specifications
  • Tip Diameter (in)0.188
  • Tip Diameter (cm)0.47752
  • DescriptionUltrasonic Processor Cup Horn Microtube Holder