Cole-Parmer Ultrasonic Processor Cup Horn, 2-1/2" Dia
Representative image only.
Cole-Parmer Ultrasonic Processor Cup Horn, 2-1/2" Dia
Cole-Parmer | Item: 0471038
Process a sample in isolation from the probe
₹192,000.00INR / Each
₹192,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Ultrasonic Processor Cup Horn, 2-1/2" Dia
Cole-Parmer Ultrasonic Processor Cup Horn, 2-1/2" Dia
0471038
₹192,000.00INR / Each
+ =
Total Price 192000.00 ADD 2 ITEMS TO CART
Product Description
Includes
vial holder and splash shield.
Key Features
  • Cooling water can be circulated through the cup during operation
  • 3 year warranty
More About this Item
Use this cup horn to process a sample in absolute isolation from the probe. The horn is especially useful for processing samples that must be kept in a sealed vessel, such as pathogens. The ultrasonic vibrations are transmitted through the water in the cup causing cavitation in the sample vessel.
3 Year Warranty
Specifications
  • Tip Diameter (in)2.5
  • Tip Diameter (cm)6.35
  • DescriptionUltrasonic Processor Cup Horn, 2-1/2" Dia