Cole-Parmer Ultra High Molecular Weight tape 2" 5 yards .005" 15ft/rl

Representative image only.

Cole-Parmer Ultra High Molecular Weight tape 2" 5 yards .005" 15ft/rl

Cole-Parmer | Item: 0827724

₹5,800.00INR / Roll of 1

₹5,800.00INR / Roll of 1
Product Description

Key Features

  • 1 year warranty

More About this Item

UHMW (Ultra High Molecular Weight) polyethylene provides a slippery, high-slide, low-friction surface similar to PTFE tape but with higher abrasion and puncture resistance. UHMW tape is much tougher than PTFE and more economical. UHMW is ideal for chutes, liners, packaging lines, slides, and anywhere high-pressure sliding contact occurs. UHMW is great for sound dampening, it eliminates squeaks, rattles, and buzz caused by movement of adjacent parts. Temperature range is -40 to 225°F (-40 to 107°C). UHMW film is FDA compliant for food contact.

1 Year Warranty
Specifications
  • ColorBlue
  • Thickness (mil)5
  • MaterialPolyethylene
  • Width (in)2
  • Length (ft)15
  • Width (cm)5.1
  • Length (m)4.57
  • DescriptionUltra High Molecular Weight tape 2" 5 yards .005" 15ft/rl
  • Warranty1 year
Reviews
Q&A