Cole-Parmer Ultra Flex Lab Frame Connector, Cast Alloy
Representative image only.
Cole-Parmer Ultra Flex Lab Frame Connector, Cast Alloy
Cole-Parmer | Item: 0812942
Ultra Flex support system offers the ultimate in flexibility for laboratory use
₹20,500.00INR / Each
₹20,500.00INR / Each
Product Description
Key Features
  • Nickel-plate is corrosion-resistant and durable
  • Clamps, arms, and bases crate numerous combinations for greater versatility
  • 2 year warranty
More About this Item
Keep spares handy or expand the capabilities of your support system.
Two year warranty
Specifications
  • Clamp MaterialCast Alloy
  • Clamp TypeFrame and Rod
  • Max Grip Range (InchShort)0.75
  • Accepts Rod Diameter (in)0.75
  • DescriptionUltra Flex Lab Frame Connector, Cast Alloy