Cole-Parmer Ultra Flex 18" Flex Arm (Nickel Plated)

Representative image only.

Cole-Parmer Ultra Flex 18" Flex Arm (Nickel Plated)

Cole-Parmer | Item: 0812943

Ultra Flex support system offers the ultimate in flexibility for laboratory use

₹75,400.00INR / Each

₹75,400.00INR / Each
Product Description

Key Features

  • Nickel-plate is corrosion-resistant and durable
  • Clamps, arms, and bases crate numerous combinations for greater versatility
  • 2 year warranty

More About this Item

Keep spares handy or expand the capabilities of your support system.

Two year warranty
Specifications
  • Rod MaterialNickel-plated Zinc
  • Length (in)18
  • DescriptionUltra Flex 18" Flex Arm (Nickel Plated)
  • Warranty2 year
Reviews
Q&A