Cole-Parmer Two-Stage Low-Pressure Specialty Regulator; CGA 350
Representative image only.
Cole-Parmer Two-Stage Low-Pressure Specialty Regulator; CGA 350
Cole-Parmer | Item: 0327420
₹332,000.00INR / Each
₹332,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Model 03274-10 is a single-stage unit specifically designed for gases that attack brass or copper-bearing alloys. When used with low-vapor-pressure liquefied gases, no inlet gauge is needed. Rubber-supported PTFE diaphragm, KEL-F seat, aluminum-alloy body and bonnet, SS delivery gauge, and 10 mm sintered SS filter.
1 Year Warranty
Specifications
  • Max Delivery Pressure (psig)25
  • Max Inlet Pressure (PSI)3000
  • Outlet Connections¼" NPT(M)
  • CGA Fitting350
  • Capacity (SCFH)240
  • Outlet Gauge Graduations (PSI)1
  • Inlet Gauge Graduations (PSI)50
  • DescriptionTwo-Stage Low-Pressure Specialty Regulator; CGA 350