Cole-Parmer Two-Stage High-Purity Gas Regulator, 900 scfh Capacity, 350 CGA
Representative image only.
Cole-Parmer Two-Stage High-Purity Gas Regulator, 900 scfh Capacity, 350 CGA
Cole-Parmer | Item: 9821012
₹183,000.00INR / Each
₹183,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Ideal for corrosive or noncorrosive gas applications. Designed and constructed for diffusion resistance and easy cleanup. Only brass, stainless steel (SS), PTFE, and Kel-F® contact the operating media. Metal diaphragm, packless valves, and high-purity seals minimize or eliminate outgoing and inboard diffusion. Supplied with SS screen filter (25 µm); and brass body, bonnet, and gauges. Two-Stage High-Purity Regulators let you precisely regulate working pressure.
1 Year Warranty
Specifications
  • Max Delivery Pressure (psig)100
  • Max Inlet Pressure (PSI)3000
  • Outlet Connections¼" NPT(F)
  • CGA Fitting350
  • Capacity (SCFH)900
  • Outlet Gauge Graduations (PSI)5
  • Inlet Gauge Graduations (PSI)100
  • DescriptionTwo-Stage High-Purity Gas Regulator, 900 scfh Capacity, 350 CGA