Cole-Parmer Two-Prong Extension Clamp, Single, Large
Representative image only.
Cole-Parmer Two-Prong Extension Clamp, Single, Large
Cole-Parmer | Item: 0802144
Securely hold your flasks, beakers and burettes
₹7,900.00INR / Each
₹7,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Two-Prong Extension Clamp, Single, Large
Cole-Parmer Two-Prong Extension Clamp, Single, Large
0802144
₹7,900.00INR / Each
+
Cole-Parmer Replacement Sleeves, Red Vinyl, Large, 2
0802782
₹1,900.00INR / Each
=
Total Price 9800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
  • 1 year warranty
More About this Item
Ideal for holding straight-sided apparatus such as burets, thermometers, beakers, and flask necks. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range. Available in medium and large sizes. Dual adjustable clamps are offered in either stainless steel or nickel plated zinc. Supplied with non-slip vinyl sleeves or for temperatures above 212°F (100°C), fiberglass covers.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc with red vinyl and black fiberglass sleeves
  • Clamp TypeEquipment, Two- or Three-Prong
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)3.625
  • Length (in)9.031
  • Depth (in)0.43
  • DescriptionTwo-Prong Extension Clamp, Single, Large