Cole-Parmer Tungsten Lamp for TSP UV100/UV1000/2000
Representative image only.
Cole-Parmer Tungsten Lamp for TSP UV100/UV1000/2000
Cole-Parmer | Item: 1195837
Economical replacement tungsten lamps save you money
₹71,400.00INR / Each
₹71,400.00INR / Each
Product Description
Key Features
  • Ideal replacement for UV and visible HPLC detector lamps
  • Manufactured in an ISO-accredited Facility
  • 6 month warranty
More About this Item
Our Kinesis® range of tungsten lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
Please Note: Vials, caps, and septa formerly under the Kinesis brand have now been rebranded Cole-Parmer. All the same great features and specifications, just with a new and improved look.
6 Month Warranty
Specifications
  • Compatible WithTSP UV100/UV1000/2000
  • DescriptionTungsten Lamp for TSP UV100/UV1000/2000
  • Warranty6 Months