Cole-Parmer Tuff-Tip pH Electrode 100 Ohm PT RTD(Mini Audio Plug), BNC, 3/4"
Representative image only.
Cole-Parmer Tuff-Tip pH Electrode 100 Ohm PT RTD(Mini Audio Plug), BNC, 3/4"
Cole-Parmer | Item: 2702002
Less chance of breakage
₹49,200.00INR / Each
₹49,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Tuff-Tip pH Electrode 100 Ohm PT RTD(Mini Audio Plug), BNC, 3/4"
Cole-Parmer Tuff-Tip pH Electrode 100 Ohm PT RTD(Mini Audio Plug), BNC, 3/4"
2702002
₹49,200.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ =
Total Price 178100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Don't worry about bulb breakage
  • Eliminate electrical noise with graphite-tipped models
  • 1 year warranty
More About this Item
The unique Tuff-Tip design helps protect bulbs. Choose chemical-resistant Ryton® PPS standard electrode or with graphite tip. Ideal for viscous or severe applications, these electrodes reduce contamination and clogging with uniform pore size.
Keep a Spare1 Year Warranty
Specifications
  • Electrode Body MaterialKynar®
  • Reference JunctionCeramic
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • Thread Size NPT(M) (in)3/4
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • DescriptionTuff-Tip pH Electrode 100 Ohm PT RTD(Mini Audio Plug), BNC, 3/4"