Cole-Parmer TRS5X8-S Tripod Bunsen Burner Stand, Cast Iron, 4 1/4" Ring, 9" Height
Representative image only.
Cole-Parmer TRS5X8-S Tripod Bunsen Burner Stand, Cast Iron, 4 1/4" Ring, 9" Height
Cole-Parmer | Item: 3630058
Legs can be removed for easy storage
₹2,300.00INR / Each
₹2,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer TRS5X8-S Tripod Bunsen Burner Stand, Cast Iron, 4 1/4" Ring, 9" Height
Cole-Parmer TRS5X8-S Tripod Bunsen Burner Stand, Cast Iron, 4 1/4" Ring, 9" Height
3630058
₹2,300.00INR / Each
+ =
Total Price 6300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily position over burners Angled legs provide excellent stability
  • 1 year warranty
More About this Item
These single-ring tripod stands feature a cast-iron ring with black enamel finish and detachable zinc-plated legs. Ideal for supporting flat-bottom glassware over your bunsen burner.
1 Year Warranty
Specifications
  • Height (in)9
  • DescriptionTripod Bunsen Burner Stand, Cast Iron, 4 1/4" Ring